Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016388.hg.1 |
TC01001033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016388.hg.1 |
ENST00000349139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016388.hg.1 |
ENST00000369441.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016388.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016389.hg.1 |
TC01001033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016389.hg.1 |
ENST00000349139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016389.hg.1 |
ENST00000369441.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01016389.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168964.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168964.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168964.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168964.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168964.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168964.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168964.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168964.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168964.hg.1 |
TC0100009580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100168965.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168965.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168965.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168965.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168965.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100168965.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100168965.hg.1 |
TC0100009580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000349139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000369441.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000471680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
ENST00000487202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009580.hg.1 |
NM_006784.2 |
Refseq |