Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000621314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000621314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000621314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007663.hg.1 |
ENST00000621314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
ENST00000293670.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012793.hg.1 |
NM_002282.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000620027.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000621314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000621314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000621314.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007663.hg.1 |
ENST00000621314.1 |
Ensembl |