Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016287.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016287.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016287.hg.1 |
TC12001242.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016285.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016285.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016285.hg.1 |
TC12001242.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000541175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000541175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000541175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000610639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
ENST00000610639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
NM_001097643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
NM_001097643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
NM_001097643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012763.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202688.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202688.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202688.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202688.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202688.hg.1 |
TC1200012763.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200202687.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202687.hg.1 |
ENST00000610639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202687.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202687.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202687.hg.1 |
ENST00000610639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202687.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202687.hg.1 |
TC1200012763.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200202689.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202689.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202689.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202689.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202689.hg.1 |
TC1200012763.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000531678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000539585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000541175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000541175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000541175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000610639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
ENST00000610639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
NM_001097643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
NM_001097643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
NM_001097643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
NM_176884.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012763.hg.1 |
NM_176884.2 |
Refseq |