Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000558.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000558.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000558.hg.1 |
TC20000033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000557.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000557.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000557.hg.1 |
TC20000033.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000144921.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144921.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144921.hg.1 |
TC2000006535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144921.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144921.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006535.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144920.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144920.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144920.hg.1 |
TC2000006535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144920.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144920.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144923.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144923.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144923.hg.1 |
TC2000006535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144923.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144923.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144919.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144919.hg.1 |
NM_000915.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144919.hg.1 |
TC2000006535.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144919.hg.1 |
ENST00000217386.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144919.hg.1 |
NM_000915.3 |
Refseq |