Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009214.hg.1 |
ENST00000390310.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009214.hg.1 |
ENST00000390310.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009214.hg.1 |
ENST00000579613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009214.hg.1 |
ENST00000579613.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006795.hg.1 |
ENST00000385099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006795.hg.1 |
ENST00000385099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006795.hg.1 |
ENST00000520681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006795.hg.1 |
ENST00000520681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009216.hg.1 |
ENST00000385098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009216.hg.1 |
ENST00000390314.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009216.hg.1 |
ENST00000390314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009216.hg.1 |
ENST00000385098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009216.hg.1 |
ENST00000390314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009216.hg.1 |
ENST00000390314.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006795.hg.1 |
ENST00000385099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006795.hg.1 |
ENST00000385099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006795.hg.1 |
ENST00000520681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006795.hg.1 |
ENST00000520681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009214.hg.1 |
ENST00000390310.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009214.hg.1 |
ENST00000390310.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009214.hg.1 |
ENST00000579613.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009214.hg.1 |
ENST00000579613.1 |
Ensembl |