Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010567.hg.1 |
ENST00000404155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010567.hg.1 |
ENST00000404155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010567.hg.1 |
ENST00000404155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000333628.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
ENST00000489942.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001069.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010565.hg.1 |
NM_001310315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010567.hg.1 |
ENST00000404155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010567.hg.1 |
ENST00000404155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010567.hg.1 |
ENST00000404155.1 |
Ensembl |