Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027785.hg.1 |
TC02001723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027785.hg.1 |
ENST00000342905.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027785.hg.1 |
ENST00000360906.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027785.hg.1 |
ENST00000402280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027785.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027785.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027785.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027785.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012189.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
ENST00000342905.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
ENST00000360906.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
ENST00000402280.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
ENST00000404025.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
NM_001199138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
NM_001199139.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
NM_001302504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
NM_021209.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189013.hg.1 |
TC0200012189.hg.1 |
Clariom D TC |