Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184870.1 | NP_001171799.1 | 2 - 3 | 391 | 110 | |
GenBank mRNA | AF329494.1 | - | 2 - 3 | 98 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |