Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027029.hg.1 |
ENST00000282251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027029.hg.1 |
ENST00000433523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027029.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027029.hg.1 |
TC11002259.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000417747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000462890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000462890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000462890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000462890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193877.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193877.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193877.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193877.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193877.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193877.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193877.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193877.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193877.hg.1 |
TC1100012208.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000282251.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000417747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000431778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000462890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000462890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000462890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000462890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
ENST00000532251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012208.hg.1 |
NM_152434.2 |
Refseq |