Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000257504.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000257504.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000257504.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000257504.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000257504.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000257504.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000257504.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000257504.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000257504.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388711.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000388712.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000464314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000464314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000464314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000464314.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000466178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000466178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000466178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000466178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000466178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000470762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000470762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000470762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000470762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000470762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000470762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000472919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000472919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000472919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000472919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000472919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000486130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000486130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000486130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000486130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000486130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
ENST00000486130.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_016548.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010614.hg.1 |
NM_177937.2 |
Refseq |