Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244710.1 | NP_001231639.1 | 9 - 10 | 919 | 91 | |
XM_017003801.1 | XP_016859290.1 | 9 - 10 | 883 | 91 | ||
GenBank mRNA | AF334737.1 | - | 5 - 6 | 367 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |