Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166588.2 | NP_001160060.1 | 8 - 9 | 1210 | 63 | |
NM_001261418.1 | NP_001248347.1 | 8 - 9 | 1210 | 63 | ||
NM_020365.4 | NP_065098.1 | 8 - 9 | 1210 | 63 | ||
XM_017002745.1 | XP_016858234.1 | 8 - 9 | 1098 | 63 | ||
XM_017002746.1 | XP_016858235.1 | 9 - 10 | 1177 | 63 | ||
XM_017002747.1 | XP_016858236.1 | 9 - 10 | 1171 | 63 | ||
GenBank mRNA | AF257077.1 | - | 8 - 9 | 1073 | 63 | |
AK022399.1 | - | 8 - 9 | 1072 | 63 | ||
AK024006.1 | - | 8 - 9 | 1098 | 63 | ||
AK314668.1 | - | 8 - 9 | 1083 | 63 | ||
AL834288.1 | - | 8 - 9 | 1075 | 63 | ||
AW972575.1 | - | 2 - 3 | 63 | |||
BC018728.1 | - | 8 - 9 | 1068 | 4849789 | 63 | |
BC021017.1 | - | 8 - 9 | 1061 | 63 | ||
CR457285.1 | - | 4 - 5 | 464 | 63 | ||
CX781577.1 | - | 2 - 3 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |