Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033225.5 | NP_150094.5 | 58 - 59 | 9485 | 84 | |
XM_011534752.2 | XP_011533054.1 | 58 - 59 | 9714 | 84 | ||
XM_011534753.2 | XP_011533055.1 | 41 - 42 | 6592 | 84 | ||
XM_011534754.1 | XP_011533056.1 | 34 - 35 | 5328 | 84 | ||
XM_017013731.1 | XP_016869220.1 | 58 - 59 | 9714 | 84 | ||
GenBank mRNA | AB209502.1 | - | 45 - 46 | 7348 | 84 | |
AF333704.2 | - | 59 - 60 | 9491 | 84 | ||
AK127129.1 | - | 9 - 10 | 1503 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |