Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013951.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013951.hg.1 |
ENST00000335351.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013951.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013951.hg.1 |
TC20001020.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013953.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013953.hg.1 |
ENST00000335351.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013953.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013953.hg.1 |
TC20001020.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013950.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013950.hg.1 |
ENST00000335351.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013950.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013950.hg.1 |
TC20001020.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013948.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013948.hg.1 |
ENST00000335351.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013948.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013948.hg.1 |
TC20001020.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009780.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168200.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168200.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168200.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168200.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168200.hg.1 |
TC2000009780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168200.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168200.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168200.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168200.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168203.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168203.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168203.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168203.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168203.hg.1 |
TC2000009780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168203.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168203.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168203.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168203.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168205.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168205.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168205.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168205.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168205.hg.1 |
TC2000009780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168205.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168205.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168205.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168205.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168202.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168202.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168202.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168202.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168202.hg.1 |
TC2000009780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168202.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168202.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168202.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168202.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168204.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168204.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168204.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168204.hg.1 |
NM_006602.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168204.hg.1 |
TC2000009780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168204.hg.1 |
ENST00000217162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168204.hg.1 |
ENST00000335351.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168204.hg.1 |
NM_001301726.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168204.hg.1 |
NM_006602.3 |
Refseq |