Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013845.hg.1 |
ENST00000307149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013845.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013845.hg.1 |
TC16000960.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000562842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000562842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000569635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
ENST00000569635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600174931.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600174931.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600174931.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600174931.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600174931.hg.1 |
TC1600009731.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000562842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000562842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000565747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007223.hg.1 |
ENST00000570080.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000307149.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000561854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000563164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000566364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000567821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000569635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
ENST00000569635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009731.hg.1 |
NM_153603.3 |
Refseq |