Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008758.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008758.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008758.hg.1 |
TC20000602.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158616.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158616.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158616.hg.1 |
TC2000008316.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158616.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158616.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
ENST00000378858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008316.hg.1 |
NM_152611.4 |
Refseq |