Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003691.hg.1 |
ENST00000263182.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003691.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003691.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003691.hg.1 |
ENST00000528583.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003691.hg.1 |
ENST00000529202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003691.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003691.hg.1 |
TC11000278.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000534323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000534323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000534323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
ENST00000534323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150150.hg.1 |
TC1100007111.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000263182.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000525090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000527505.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000528583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000529202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000531832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000533566.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000534323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000534323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000534323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
ENST00000534323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007111.hg.1 |
NM_003986.2 |
Refseq |