Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004460.hg.1 |
TC02000293.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004460.hg.1 |
ENST00000263735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004460.hg.1 |
ENST00000405271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004460.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004462.hg.1 |
TC02000293.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004462.hg.1 |
ENST00000263735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004462.hg.1 |
ENST00000405271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004462.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000419334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000419334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000419334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000419334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151925.hg.1 |
TC0200007506.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200151927.hg.1 |
TC0200007506.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000263735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000405271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000419334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000419334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000419334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000419334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000456133.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000474691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
ENST00000490733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007506.hg.1 |
NM_002354.2 |
Refseq |