Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000221.hg.1 |
TC17_ctg5_hap1000017.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000221.hg.1 |
NM_001123066.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000221.hg.1 |
NM_001123067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000221.hg.1 |
NM_001203251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000221.hg.1 |
NM_001203252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000221.hg.1 |
NM_005910.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000221.hg.1 |
NM_016834.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000221.hg.1 |
NM_016835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000221.hg.1 |
NM_016841.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000262410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000334239.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000340799.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000344290.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000347967.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000351559.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000354326.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000415613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000420682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000446361.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
ENST00000535772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
NM_001123066.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
NM_001123067.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
NM_001203251.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
NM_001203252.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
NM_005910.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
NM_016834.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
NM_016835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
NM_016841.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17007253.hg.1 |
TC17000592.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000262410.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000334239.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000340799.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000344290.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000351559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000415613.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000420682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000431008.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000446361.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000535772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000570299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000571987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000574436.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000576518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000577017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_001123066.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_001203252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_016834.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_016835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_016841.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000262410.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000334239.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000340799.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000344290.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000351559.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000415613.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000420682.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000431008.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000446361.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000535772.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000570299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000571987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000574436.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000576518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
ENST00000577017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_001123066.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_001203252.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_016834.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_016835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
NM_016841.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700159252.hg.1 |
TC1700008082.hg.1 |
Clariom D TC |