Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006671.hg.1 |
TC0X002321.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006671.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006671.hg.1 |
ENST00000372625.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006671.hg.1 |
ENST00000372626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006671.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006671.hg.1 |
ENST00000537029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006671.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006671.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006671.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006669.hg.1 |
TC0X002321.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006669.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006669.hg.1 |
ENST00000372625.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006669.hg.1 |
ENST00000372626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006669.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006669.hg.1 |
ENST00000537029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006669.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006669.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006669.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006673.hg.1 |
TC0X002321.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006673.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006673.hg.1 |
ENST00000372625.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006673.hg.1 |
ENST00000372626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006673.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006673.hg.1 |
ENST00000537029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006673.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006673.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006673.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006668.hg.1 |
TC0X002321.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006668.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006668.hg.1 |
ENST00000372625.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006668.hg.1 |
ENST00000372626.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006668.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006668.hg.1 |
ENST00000537029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006668.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006668.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006668.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011317.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177855.hg.1 |
TC0X00011317.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
ENST00000372624.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
ENST00000372625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
ENST00000372626.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
ENST00000469820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
NM_001006639.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
NM_001006640.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
NM_004780.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177856.hg.1 |
TC0X00011317.hg.1 |
Clariom D TC |