Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006724841.3 | XP_006724904.1 | 3 - 4 | 365 | 78 | |
XM_006724842.3 | XP_006724905.1 | 3 - 4 | 365 | 78 | ||
XM_017029765.1 | XP_016885254.1 | 3 - 4 | 812 | 78 | ||
GenBank mRNA | AL531401.3 | - | 3 - 4 | 700 | 78 | |
KU178351.1 | - | 3 - 4 | 374 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |