Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021078.2 | NP_066564.2 | 15 - 16 | 2235 | 103 | |
XM_006721817.3 | XP_006721880.1 | 15 - 16 | 2267 | 103 | ||
XM_006721818.3 | XP_006721881.1 | 15 - 16 | 2150 | 103 | ||
XM_017024448.1 | XP_016879937.1 | 15 - 16 | 2398 | 103 | ||
GenBank mRNA | AF029777.1 | - | 15 - 16 | 2199 | 103 | |
BC032743.1 | - | 15 - 16 | 2212 | 5575574 | 103 | |
BC039907.1 | - | 15 - 16 | 2227 | 6064838 | 103 | |
BC105977.1 | - | 15 - 16 | 2204 | 6137532 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |