Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000420074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000420074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000420074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000588753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000588753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000588753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000588753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000590829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000590829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000590829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000590829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
TC2100007460.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150592.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
TC2100007460.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150593.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000414659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000420074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000420074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000420074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000421927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000432735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000444998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000455567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000588753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000588753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000588753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000588753.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000590829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000590829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000590829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000590829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
ENST00000591223.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110566.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007460.hg.1 |
NR_110567.1 |
Refseq |