Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000535685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000535685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000535685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000535685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012111.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012111.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012111.hg.1 |
TC12000933.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200167224.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167224.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167224.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167224.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167224.hg.1 |
TC1200009096.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000327554.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000535685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000535685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000535685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000535685.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000536742.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000539847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
ENST00000546345.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009096.hg.1 |
NM_178499.4 |
Refseq |