Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005089.hg.1 |
ENST00000372622.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005089.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005089.hg.1 |
TC20000356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
TC2000007502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152245.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000372622.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000415790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000435014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000449078.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000456939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
ENST00000467701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007502.hg.1 |
NM_052951.2 |
Refseq |