Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000359801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000381630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000381630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000381630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000381630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000381630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000381630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000381630.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000444444.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000481731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000486775.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000608747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000609796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000609796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000609796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000609796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000609796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000609796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
ENST00000609796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001122962.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009968.hg.1 |
NM_001134836.1 |
Refseq |