Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000588132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000588132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000588132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
ENST00000588132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001725.hg.1 |
ENST00000261326.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001725.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001725.hg.1 |
TC18000149.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000588132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000588132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000588132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
ENST00000588132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007092.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800148725.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148725.hg.1 |
NM_017947.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800148725.hg.1 |
TC1800007092.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800148725.hg.1 |
ENST00000261326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148725.hg.1 |
NM_017947.3 |
Refseq |