Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000416284.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000416284.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000416284.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000416284.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000416284.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000416284.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000416284.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000416284.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000416284.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000500610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000500610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000500610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000500610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000500610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000547941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000548541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000548541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000548541.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000548780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000548780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000548780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000548780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549379.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549456.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549456.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549456.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549456.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549456.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549456.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549456.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549958.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549958.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000549958.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000550396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000551619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000552075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000552075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000552075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000552075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000552075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
ENST00000552075.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
NM_178539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
NM_178539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
NM_178539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
NM_178539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
NM_178539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
NM_178539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
NM_178539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
NM_178539.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011037.hg.1 |
NM_178539.4 |
Refseq |