Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009900.hg.1 |
ENST00000221403.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009900.hg.1 |
ENST00000522614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009900.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009900.hg.1 |
TC19000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165460.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165460.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165460.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165460.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165460.hg.1 |
TC1900008503.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165460.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165460.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165460.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165460.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000221403.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000520557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000522614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
ENST00000523250.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008503.hg.1 |
NM_014475.3 |
Refseq |