Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000478830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000478830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000478830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000478830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010092.hg.1 |
TC03000549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010092.hg.1 |
ENST00000295756.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010092.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03010092.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000478830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000478830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000478830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000478830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000484927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
ENST00000493604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008290.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158987.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158987.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158987.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158987.hg.1 |
ENST00000295756.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158987.hg.1 |
ENST00000426646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300158987.hg.1 |
NM_016388.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300158987.hg.1 |
TC0300008290.hg.1 |
Clariom D TC |