Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000497381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000497381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
ENST00000497381.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020370.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020370.hg.1 |
ENST00000370483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020370.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020370.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10020370.hg.1 |
TC10001584.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000497381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000497381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
ENST00000497381.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011592.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179032.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179032.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179032.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179032.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179032.hg.1 |
ENST00000016171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179032.hg.1 |
ENST00000370483.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000179032.hg.1 |
NM_004376.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179032.hg.1 |
NM_078470.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000179032.hg.1 |
TC1000011592.hg.1 |
Clariom D TC |