Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002838.3 | NP_001002838.1 | 15 - 16 | 3015 | 94 | |
NM_020922.4 | NP_065973.2 | 15 - 16 | 3015 | 94 | ||
XM_006724593.3 | XP_006724656.1 | 15 - 16 | 2953 | 94 | ||
XM_011530802.2 | XP_011529104.1 | 15 - 16 | 2953 | 94 | ||
XM_017029741.1 | XP_016885230.1 | 15 - 16 | 2954 | 94 | ||
XM_017029742.1 | XP_016885231.1 | 15 - 16 | 3031 | 94 | ||
XM_017029743.1 | XP_016885232.1 | 15 - 16 | 2946 | 94 | ||
XM_017029744.1 | XP_016885233.1 | 15 - 16 | 2848 | 94 | ||
XM_017029745.1 | XP_016885234.1 | 15 - 16 | 2953 | 94 | ||
GenBank mRNA | AB046786.1 | - | 9 - 10 | 1287 | 94 | |
AJ409088.2 | - | 14 - 15 | 2576 | 94 | ||
AY082340.1 | - | 14 - 15 | 2576 | 94 | ||
AY352048.1 | - | 15 - 16 | 3015 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Homo sapiens Probe spans exons |