Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010447.hg.1 |
ENST00000332271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010447.hg.1 |
ENST00000527701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010447.hg.1 |
ENST00000532537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010447.hg.1 |
ENST00000533313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010447.hg.1 |
NM_001008496.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010447.hg.1 |
NM_001255975.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010447.hg.1 |
NR_045648.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010447.hg.1 |
NR_045649.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010447.hg.1 |
TC22000593.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
TC2200008234.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000332271.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000527701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000532537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000533313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000616349.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
NM_001008496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
NM_001255975.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
NR_045648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
NR_045649.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000332271.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000527701.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000532537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000533313.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
ENST00000616349.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
NM_001008496.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
NM_001255975.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
NR_045648.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200159033.hg.1 |
NR_045649.1 |
Refseq |