Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011670.hg.1 |
TC0X000874.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011670.hg.1 |
ENST00000344384.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011670.hg.1 |
ENST00000380470.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011670.hg.1 |
ENST00000480796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011670.hg.1 |
ENST00000537676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011670.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011670.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011670.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162555.hg.1 |
TC0X00009123.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000344384.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000380470.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000480796.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
ENST00000485437.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001012428.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_001201583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009123.hg.1 |
NM_080873.2 |
Refseq |