Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015509.3 | NP_056324.2 | 4 - 5 | 467 | 91 | |
GenBank mRNA | AK074858.1 | - | 4 - 5 | 424 | 91 | |
AK074880.1 | - | 4 - 5 | 437 | 91 | ||
AK074923.1 | - | 4 - 5 | 410 | 91 | ||
AK075013.1 | - | 4 - 5 | 444 | 91 | ||
BC002888.2 | - | 4 - 5 | 369 | 91 | ||
BC067367.1 | - | 2 - 3 | 139 | 91 | ||
BC110876.1 | - | 4 - 5 | 401 | 5553942 | 91 | |
DA496321.1 | - | 4 - 5 | 467 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |