Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135110.2 | NP_001128582.1 | 7 - 8 | 1070 | 76 | |
NM_001286516.1 | NP_001273445.1 | 5 - 6 | 721 | 76 | ||
NM_005147.5 | NP_005138.3 | 7 - 8 | 1070 | 76 | ||
GenBank mRNA | AB209923.1 | - | 7 - 8 | 989 | 76 | |
AF061749.1 | - | 7 - 8 | 1024 | 76 | ||
AF411044.1 | - | 7 - 8 | 1037 | 76 | ||
AK001333.1 | - | 7 - 8 | 1010 | 76 | ||
AK223105.1 | - | 7 - 8 | 1010 | 76 | ||
AK295391.1 | - | 5 - 6 | 661 | 76 | ||
AK314218.1 | - | 7 - 8 | 1007 | 76 | ||
BC007225.1 | - | 7 - 8 | 990 | 76 | ||
BC011855.2 | - | 7 - 8 | 994 | 4139881 | 76 | |
BC012343.1 | - | 7 - 8 | 986 | 4653507 | 76 | |
BC014062.2 | - | 7 - 8 | 986 | 76 | ||
BC020248.1 | - | 3 - 4 | 319 | 76 | ||
BC030145.2 | - | 7 - 8 | 986 | 76 | ||
BC032100.2 | - | 7 - 8 | 1008 | 4660602 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |