Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000509309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000509309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000509309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000509309.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016537.hg.1 |
TC04001208.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016537.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016537.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400173042.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173042.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173042.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400173042.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173042.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400173042.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400173042.hg.1 |
TC0400010733.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000263923.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000509309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000509309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000509309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000509309.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
ENST00000512566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010733.hg.1 |
NM_002253.2 |
Refseq |