Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004108.hg.1 |
TC03000207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004108.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004108.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004104.hg.1 |
TC03000207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004104.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004104.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004103.hg.1 |
TC03000207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004103.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004103.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004106.hg.1 |
TC03000207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004106.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004106.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004109.hg.1 |
TC03000207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004109.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004109.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149987.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149987.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149987.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149987.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149987.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149987.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149987.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149987.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149987.hg.1 |
TC0300007115.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149980.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149980.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149980.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149980.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149980.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149980.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149980.hg.1 |
TC0300007115.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149981.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149981.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149981.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149981.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149981.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149981.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149981.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149981.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149981.hg.1 |
TC0300007115.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149984.hg.1 |
TC0300007115.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149982.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149982.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149982.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149982.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149982.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149982.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149982.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149982.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149982.hg.1 |
TC0300007115.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149986.hg.1 |
TC0300007115.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000232905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000462088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000487151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
ENST00000488260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007115.hg.1 |
NM_005875.2 |
Refseq |