Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007880.hg.1 |
ENST00000265992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007880.hg.1 |
ENST00000403870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007880.hg.1 |
ENST00000419934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007880.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007880.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007880.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007880.hg.1 |
TC10000679.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011169.hg.1 |
ENST00000450600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009045.hg.1 |
ENST00000457810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007882.hg.1 |
ENST00000265992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007882.hg.1 |
ENST00000403870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007882.hg.1 |
ENST00000419934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007882.hg.1 |
ENST00000534974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007882.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007882.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007882.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007882.hg.1 |
TC10000679.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007881.hg.1 |
ENST00000265992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007881.hg.1 |
ENST00000403870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007881.hg.1 |
ENST00000419934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007881.hg.1 |
ENST00000534974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007881.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007881.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007881.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007881.hg.1 |
TC10000679.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008529.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009045.hg.1 |
ENST00000457810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158164.hg.1 |
TC1000008529.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158165.hg.1 |
TC1000008529.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
ENST00000465148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158166.hg.1 |
TC1000008529.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011169.hg.1 |
ENST00000450600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
ENST00000265992.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
ENST00000403870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
NM_001134375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
NM_001134376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
NM_019084.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158163.hg.1 |
TC1000008529.hg.1 |
Clariom D TC |