Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025904.hg.1 |
TC06002071.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025904.hg.1 |
ENST00000339697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025904.hg.1 |
ENST00000368454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025904.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190065.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600190065.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190065.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600190065.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600190065.hg.1 |
TC0600013054.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
ENST00000339697.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013054.hg.1 |
NM_020755.3 |
Refseq |