Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031952.3 | NP_114158.2 | 2 - 3 | 301 | 104 | |
NR_125327.1 | 2 - 3 | 301 | 104 | |||
NR_125328.1 | 2 - 3 | 276 | 104 | |||
NR_125329.1 | 2 - 3 | 301 | 104 | |||
XM_011543664.1 | XP_011541966.1 | 2 - 3 | 257 | 104 | ||
XM_017009948.1 | XP_016865437.1 | 2 - 3 | 282 | 104 | ||
XM_017009949.1 | XP_016865438.1 | 7 - 8 | 838 | 104 | ||
XM_017009950.1 | XP_016865439.1 | 6 - 7 | 820 | 104 | ||
GenBank mRNA | AK292338.1 | - | 2 - 3 | 282 | 104 | |
AK310260.1 | - | 2 - 3 | 257 | 104 | ||
AY027526.1 | - | 2 - 3 | 250 | 104 | ||
BC032832.1 | - | 2 - 3 | 306 | 4827641 | 104 | |
BC047333.1 | - | 2 - 3 | 287 | 5166271 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |