Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012079.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012079.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12012079.hg.1 |
TC12000930.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000537597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
ENST00000545224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009085.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167156.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167156.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167156.hg.1 |
ENST00000267260.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200167156.hg.1 |
NM_194286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200167156.hg.1 |
TC1200009085.hg.1 |
Clariom D TC |