Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPV17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BG714998.1 | - | 5 - 6 | 329 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |