Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001385.hg.1 |
TC02000131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001385.hg.1 |
ENST00000486442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001385.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147253.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147253.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147253.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147253.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147253.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147253.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147253.hg.1 |
TC0200006936.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000288548.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000471654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000486442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
ENST00000489446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006936.hg.1 |
NM_052920.1 |
Refseq |