Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011518843.1 | XP_011517145.1 | 1 | 871 | 135 | |
GenBank mRNA | AL353935.1 | - | 1 | 256 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |