Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049661.hg.1 |
TC01003225.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049661.hg.1 |
ENST00000290524.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049661.hg.1 |
ENST00000368870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049661.hg.1 |
ENST00000452513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049661.hg.1 |
ENST00000452671.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049661.hg.1 |
ENST00000478564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049661.hg.1 |
NM_000449.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01049661.hg.1 |
NM_001025603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000290524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000368870.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000392746.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000412774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000421986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000422595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000430227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000435314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000436271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000437327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000444392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000450506.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000452456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000452671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000458484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000478564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000479681.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000494217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
NM_000449.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
NM_001025603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000290524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000368870.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000392746.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000412774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000421986.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000422595.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000430227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000435314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000436271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000437327.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000444392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000450506.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000452456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000452671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000458484.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000478564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000479681.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
ENST00000494217.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
NM_000449.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
NM_001025603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100218460.hg.1 |
TC0100015783.hg.1 |
Clariom D TC |