Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007434.hg.1 |
TC07000523.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007434.hg.1 |
ENST00000338056.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007434.hg.1 |
ENST00000394654.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007434.hg.1 |
ENST00000493037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007434.hg.1 |
NM_001134405.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007434.hg.1 |
NM_001134406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007434.hg.1 |
NM_138290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000312373.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000338056.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000394654.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000466676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000476114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000489461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000493037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000497788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
NM_001134405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
NM_001134406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
NM_138290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000312373.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000338056.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000394654.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000466676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000476114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000489461.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000493037.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
ENST00000497788.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
NM_001134405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
NM_001134406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
NM_138290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157355.hg.1 |
TC0700008261.hg.1 |
Clariom D TC |