Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014368.hg.1 |
TC03000800.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014368.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014368.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014366.hg.1 |
TC03000800.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014366.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014366.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014367.hg.1 |
TC03000800.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014367.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014367.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
ENST00000477926.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009122.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165384.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165384.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165384.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165384.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165384.hg.1 |
TC0300009122.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300165386.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165386.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165386.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165386.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165386.hg.1 |
TC0300009122.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300165387.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165387.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165387.hg.1 |
ENST00000296046.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165387.hg.1 |
NM_001870.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165387.hg.1 |
TC0300009122.hg.1 |
Clariom D TC |