Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000677.hg.1 |
TC12003206.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000677.hg.1 |
ENST00000252318.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000677.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000535354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000535354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
ENST00000535354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200198797.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200198797.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200198797.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200198797.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200198797.hg.1 |
TC1200012577.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000252318.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000535354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000535354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
ENST00000535354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012577.hg.1 |
NM_017417.1 |
Refseq |