Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007488.hg.1 |
TC09000649.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007488.hg.1 |
ENST00000297933.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007488.hg.1 |
ENST00000312123.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007488.hg.1 |
ENST00000394083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007488.hg.1 |
ENST00000394104.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007488.hg.1 |
ENST00000394105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007488.hg.1 |
ENST00000470056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007488.hg.1 |
ENST00000495955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007488.hg.1 |
NM_015635.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
TC09000649.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
ENST00000265956.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
ENST00000297933.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
ENST00000312123.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
ENST00000394083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
ENST00000394104.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
ENST00000394105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
ENST00000470056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
ENST00000495955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007489.hg.1 |
NM_015635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000265956.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000297933.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000312123.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000394083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000394104.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000394105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000436712.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000467750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000470056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000479511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000495955.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000497580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
NM_001282679.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
NM_001282680.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
NM_001282681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
NM_015635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000265956.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000297933.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000312123.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000394083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000394104.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000394105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000436712.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000467750.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000470056.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000479511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000495955.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
ENST00000497580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
NM_001282679.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
NM_001282680.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
NM_001282681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
NM_015635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158393.hg.1 |
TC0900008750.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900158390.hg.1 |
ENST00000394105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158390.hg.1 |
NM_015635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158390.hg.1 |
ENST00000394105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158390.hg.1 |
NM_015635.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158390.hg.1 |
TC0900008750.hg.1 |
Clariom D TC |